1-Chloro-3,4-dinitrobenzene 90%
Supplier: Thermo Scientific Chemicals
|
Danger
|
172311000
172312500
172310500
TS172311000EA
74
USD
TS172311000
TS172312500
TS172310500
1-Chloro-3,4-dinitrobenzene 90%
1-Chloro-3,4-dinitrobenzene
Formula:
C₆H₃ClN₂O₄ Boiling Pt: 315 °C (760 mmHg) Melting Pt: 36 °C Density: 1.68 g/cm³ Flash Pt: 194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007212 CAS Number: 610-40-2 EINECS: 210-223-7 UN: 3441 ADR: 6.1,II |
Specification Test Results
Appearance | Yellow-brown Viscous liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.5865 to 1.5885 (20 °C, 589 nm) |
GC | ≥88 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...