Dimethallyl carbonate 97%

Supplier: Thermo Scientific Chemicals

170580050 170580250
TS170580050EA 43 USD
TS170580050 TS170580250
Dimethallyl carbonate 97%
Dimethallyl carbonate
Formula: C₉H₁₄O₃
Boiling Pt: 199…202 °C (760 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD00008598
CAS Number: 64057-79-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Loss on drying ≤0.5 % (80 °C, 4 h) (vacuum)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR