2-Chloro-3-hydroxypyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-3-pyridinol

152180250 152181000
TS152180250EA 68 USD
TS152180250 TS152181000
2-Chloro-3-hydroxypyridine 98%
2-Chloro-3-hydroxypyridine
Formula: C₅H₄ClNO
MW: 129.55 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00006235
CAS Number: 6636-78-8
EINECS: 229-635-3

Order Now

Specification Test Results

Appearance Light beige Powder
Infrared spectrum Conforms
Melting point 168 to 172 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR