N-(4-Bromobutyl)phthalimide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Bromobutyl)isoindoline-1,3-dione

146290100 146290500
TS146290100EA 103 USD
TS146290100 TS146290500
N-(4-Bromobutyl)phthalimide 98%
N-(4-Bromobutyl)phthalimide
Formula: C₁₂H₁₂BrNO₂
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00005905
CAS Number: 5394-18-3
EINECS: 226-401-2

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR