4,4'-Dichlorobutyrophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

136890250 136891000
TS136890250EA 32 USD
TS136890250 TS136891000
4,4'-Dichlorobutyrophenone 97%
4,4'-Dichlorobutyrophenone
Formula: C₁₀H₁₀Cl₂O
Melting Pt: 29…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00018993
CAS Number: 40877-09-6
EINECS: 255-123-4

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline mass or liquid (May darken after melting)
Infrared spectrum Conforms
GC ≥96.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR