2-Amino-6-chlorobenzothiazole 99%

Supplier: Thermo Scientific Chemicals
Warning

136400100 136400500
TS136400100EA 28 USD
TS136400100 TS136400500
2-Amino-6-chlorobenzothiazole 99%
2-Amino-6-chlorobenzothiazole
Formula: C₇H₅ClN₂S
Melting Pt: 197…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00053557
CAS Number: 95-24-9
EINECS: 202-402-3

Order Now

Specification Test Results

Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 197 to 201 °C
HPLC ≥98.5 %
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR