Charcoal activated, NORIT® SA 2
Supplier: Thermo Scientific Chemicals
134370025
134370250
134372500
TS134370025EA
235
USD
TS134370025
TS134370250
TS134372500
Charcoal activated, NORIT® SA 2
Charcoal activated
Formula:
C Melting Pt: 3652 °C Storage Temperature: Ambient |
MDL Number:
MFCD00133992 CAS Number: 7440-44-0 |
Specification Test Results
Appearance | Black Powder |
Loss on drying | ≤10 % |
Molasses number | ≤390 |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...