N-(Propargyloxy)phthalimide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(Prop-2-yn-1-yloxy)isoindoline-1,3-dione

131490010 131490100 131490250
TS131490010EA 25 USD
TS131490010 TS131490100 TS131490250
N-(Propargyloxy)phthalimide 98%
N-(Propargyloxy)phthalimide
Formula: C₁₁H₇NO₃
Melting Pt: 149…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00005890
CAS Number: 4616-63-1

Order Now

Specification Test Results

Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 149 to 152 °C
Titration with TBAH ≥97.5 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR