Phthalaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde

131080050 131080250 131081000 131085000
TS131080050EA 101 USD
TS131080050 TS131080250 TS131081000 TS131085000
Phthalaldehyde 98+%
Phthalaldehyde
Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 83…84 °C (1hPa)
Melting Pt: 54…57 °C
Flash Pt: 132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003335
CAS Number: 643-79-8
EINECS: 211-402-2
UN: 2923
ADR: 8 II

Order Now

Specification Test Results

Appearance Yellow Powder or crystals and/or lumps
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR