1,1-Dimethoxy-2-phenylethane 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Phenylacetaldehyde dimethyl acetal

130270050 130271000 130275000
TS130270050EA 14 USD
TS130270050 TS130271000 TS130275000
1,1-Dimethoxy-2-phenylethane 98%
1,1-Dimethoxy-2-phenylethane
Formula: C₆H₅CH₂CH(OCH₃)₂
MW: 166.22 g/mol
Boiling Pt: 219…221 °C (754 mmHg)
Melting Pt: 62…64 °C
Density: 1.004 g/cm³
Flash Pt: 83 °C
MDL Number: MFCD00008487
CAS Number: 101-48-4
EINECS: 202-945-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4930 to 1.4950 (20 °C, 589 nm)
GC ≥97.5 %
Specific gravity (25/25 °C) 1.002 to 1.006
Acid value ≤1 mg KOH/g
Volatile matter ≤5.0 % (105 °C, 3 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR