Pamoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,4'-Methylenebis(3-hydroxy-2-naphthoic acid), Embonic acid

129721000 129725000
TS129721000EA 111 USD
TS129721000 TS129725000
Pamoic acid 98+%
Pamoic acid
Formula: C₂₃H₁₆O₆
MW: 388.38 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004079
CAS Number: 130-85-8

Order Now

Specification Test Results

Appearance Yellow to yellow-green Crystalline powder
Infrared spectrum Conforms
Titration excess NaOH 98.0 to 102.0 %
GC ≥97.0 %
Stabilizer ≤500 ppm BHT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR