6-Nitroindoline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Dihydro-6-nitro-(1H)-indole

128710010 128710050
TS128710010EA 43 USD
TS128710010 TS128710050
6-Nitroindoline 98%
6-Nitroindoline
Formula: C₈H₈N₂O₂
MW: 164.16 g/mol
Melting Pt: 66…70 °C
MDL Number: MFCD00005710
CAS Number: 19727-83-4
EINECS: 243-257-6

Order Now

Specification Test Results

Appearance Orange Crystals
Infrared spectrum Conforms
Melting point 66 to 70 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR