8-Hydroxyquinoline-5-sulfonic acid monohydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

121920025 121921000 121925000
TS121920025EA 614 USD
TS121920025 TS121921000 TS121925000
8-Hydroxyquinoline-5-sulfonic acid monohydrate 98%
8-Hydroxyquinoline-5-sulphonic acid monohydrate
Formula: C₉H₇NO₄S·1H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149482
CAS Number: 283158-18-9
EINECS: 201-570-5

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Assay ≥97.5 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR