4-Hydroxy-1-methylpiperidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Methyl-4-piperidinol, 1-Methyl-4-piperidinol, 1-Methylpiperidin-4-ol

121510050 121511000 121515000
TS121510050EA 26 USD
TS121510050 TS121511000 TS121515000
4-Hydroxy-1-methylpiperidine 98%
4-Hydroxy-1-methylpiperidine
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 200
Melting Pt: 29…31 °C
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00006500
CAS Number: 106-52-5
EINECS: 203-406-8

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4760 to 1.4790 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %
Titration with HClO4 ≥97.5 %
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.2 ppm
Lead (Pb) ≤0.05 ppm
Magnesium (Mg) ≤0.1 ppm
Potassium (K) ≤0.2 ppm
Sodium (Na) ≤1 ppm
Zinc (Zn) ≤0.1 ppm
Total phosphorus (P) ≤0.5 ppm
Total silicon (Si) ≤0.02 ppm
Total sulfur (S) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR