3-Hydroxy-3-methylbutanone 92%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxy-3-methyl-2-butanone, 3-Hydroxy-3-methylbutan-2-one

121440050 121440250
TS121440050EA 56 USD
TS121440050 TS121440250
3-Hydroxy-3-methylbutanone 92%
3-Hydroxy-3-methylbutanone
Formula: C₅H₁₀O₂
MW: 102.13
Boiling Pt: 140
Density: 0.97 g/cm³
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00004460
CAS Number: 115-22-0
EINECS: 204-073-1
UN: 1224
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light red Liquid
Infrared spectrum Conforms
Refractive index 1.4140 to 1.4170 (20°C, 589 nm)
GC ≥90 %
UV at 240 nm A: ≤0.02
Residue after evaporation ≤2 ppm
Acidity ≤0.00015 meq/g
Filtered to 0.2 micrometer Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR