6-Propyl-2-thiouracil ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: Propylthiouracil, 4-Hydroxy-2-mercapto-6-propylpyrimidine, 4-Propyl-2-thiouracil, 6-Propyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

121310250 121311000
TS121310250EA 79 USD
TS121310250 TS121311000
6-Propyl-2-thiouracil ≥98%
6-Propyl-2-thiouracil

  • Bioactive

Formula: C₇H₁₀N₂OS
MW: 170.24 g/mol
Melting Pt: 218…221 °C
Flash Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006041
CAS Number: 51-52-5

Order Now

Specification Test Results

Appearance White to yellowish white Powder
Infrared spectrum Conforms
Melting point 217 to 221 °C
Titration ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR