Hexamethylenetetramine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexamethylenetetramine, Hexamine, Urotropine, Formin

120610010 120610025 120610100
TS120610010EA 53 USD
TS120610010 TS120610025 TS120610100
Hexamethylenetetramine 99%
Hexamethylenetetramine
Formula: C₆H₁₂N₄
MW: 140.19 g/mol
Melting Pt: 260 °C (subl.)
Density: 1.33 g/cm³
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00006895
CAS Number: 100-97-0
EINECS: 202-905-8
UN: 1328
ADR: 4.1,III
Merck Index: 13,05994

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Ash ≤0.1 %
Titration with HClO4 ≥98.5 %
NMR (1H-NMR) Consistent with structure
Particle size ≥70 % (> 32 µm)
Residue after ignition ≤0.05 %
pH 5 to 7

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR