2,6-Dimethylpiperazine 98%

Supplier: Thermo Scientific Chemicals
Danger

116670250 116671000
TS116670250EA 66 USD
TS116670250 TS116671000
2,6-Dimethylpiperazine 98%
2,6-Dimethylpiperazine
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 162 °C (760 mmHg)
Melting Pt: 108…113 °C
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00005956
CAS Number: 108-49-6
UN: 2925
ADR: 4.1,II

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 108 to 113 °C
Water ≤2 %
GC ≥97.5 %
Calcium (Ca) ≤0.05 %
Iron (Fe) ≤0.001 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.01 %
Strontium (Sr) ≤0.8 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 % (in dilute HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR