4,5-Dichloro-o-phenylenediamine 98%

Supplier: Thermo Scientific Chemicals
Warning

113640050 113640250
TS113640050EA 84 USD
TS113640050 TS113640250
4,5-Dichloro-o-phenylenediamine 98%
4,5-Dichloro-o-phenylenediamine
Formula: C₆H₆Cl₂N₂
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 162…163 °C
MDL Number: MFCD00007723
CAS Number: 5348-42-5
EINECS: 226-305-0

Order Now

Specification Test Results

Appearance Red-brown to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR