1,2-Dichloro-4-nitrobenzene 95%

Supplier: Thermo Scientific Chemicals
Warning

113530050 113532500
TS113530050EA 18 USD
TS113530050 TS113532500
1,2-Dichloro-4-nitrobenzene 95%
1,2-Dichloro-4-nitrobenzene
Formula: C₆H₃Cl₂NO₂
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 39…45 °C
Flash Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00007207
CAS Number: 99-54-7
EINECS: 202-764-2

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline mass
Infrared spectrum Conforms
Melting point 39 to 45 °C
GC ≥94 %
Calcium (Ca) ≤10 ppm
Chromium (Cr) ≤0.2 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.4 ppm
Lead (Pb) ≤0.2 ppm
Manganese (Mn) ≤0.1 ppm
Nickel (Ni) ≤0.2 ppm
Palladium (Pd) ≤0.1 ppm
Sulfur (S) ≤20 ppm
Zinc (Zn) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR