9,10-Dibromoanthracene 96%

Supplier: Thermo Scientific Chemicals
Warning

112710500 112710100
TS112710500EA 165 USD
TS112710500 TS112710100
9,10-Dibromoanthracene 96%
9,10-Dibromoanthracene
Formula: C₁₄H₈Br₂
Boiling Pt: 427 °C (1013 hPa)
Melting Pt: 220…225 °C
MDL Number: MFCD00001244
CAS Number: 523-27-3
EINECS: 208-342-4

Order Now

Specification Test Results

Appearance Yellow to yellow-green Powder
Infrared spectrum Conforms
Melting point 220 to 225 °C
GC ≥95 %
Iodide (I) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Sodium (Na) ≤0.005 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %
Nitrogen compounds (as N) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR