4-Cyanobenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

110730050 110730250
TS110730050EA 46 USD
TS110730050 TS110730250
4-Cyanobenzoic acid 99%
4-Cyanobenzoic acid
Formula: C₈H₅NO₂
MW: 147.13 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 217…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00002528
CAS Number: 619-65-8
EINECS: 210-606-9

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 217 to 222 °C
HPLC ≥98.5 %
Titration by coupling ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR