4-Chloro-1-naphthol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloronaphthalen-1-ol, 1-Chloro-4-hydroxynaphthalene, 4-CN, Chloronaphthol

109550050 109550250 109551000
TS109550050EA 79 USD
TS109550050 TS109550250 TS109551000
4-Chloro-1-naphthol 97%
4-Chloro-1-naphthol
Formula: ClC₁₀H₆OH
Boiling Pt: 307 °C (1013 hPa)
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00003974
CAS Number: 604-44-4
EINECS: 210-068-5

Order Now

Specification Test Results

Appearance Off-white to grey to light brown Powder or needles
Infrared spectrum Conforms
Melting point 117 to 122 °C
GC ≥96.0 %
UV at 280 nm A: ≤0.1
Titration after Ion exchange ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR