4-Bromophenyl phenyl ether (BDE No. 3) 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Bromodiphenyl ether, 1-Bromo-4-phenoxybenzene

106890050 106890250
TS106890050EA 22 USD
TS106890050 TS106890250
4-Bromophenyl phenyl ether (BDE No. 3) 99%
4-Bromophenyl phenyl ether (BDE No. 3)
Formula: C₁₂H₉BrO
Boiling Pt: 305 °C (760 mmHg)
Melting Pt: 18 °C
Density: 1.423 g/cm³
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000094
CAS Number: 101-55-3
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.6065 to 1.6095 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR