4-Benzylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

106160050 106161000
TS106160050EA 27 USD
TS106160050 TS106161000
4-Benzylpyridine 97%
4-Benzylpyridine
Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 9…11 °C
Density: 1.06 g/cm³
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00006443
CAS Number: 2116-65-6
EINECS: 218-319-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5810 to 1.5830 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR