5-Benzyloxyindole 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Benzyloxy-1H-indole, 5-(Benzyloxy)-1H-indole

106090250 106090050
TS106090250EA 148 USD
TS106090250 TS106090050
5-Benzyloxyindole 95%
5-Benzyloxyindole
Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
Boiling Pt: 165 °C (0.1 mmHg)
Melting Pt: 96…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00005676
CAS Number: 1215-59-4
EINECS: 214-930-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to beige Powder
Infrared spectrum Conforms
GC ≤6 % Toluene
UV at 235 nm A: ≤1.0
Color scale ≤10 APHA
Stabilizer Approx. 50 ppm Amylene
Residue after evaporation ≤0.001 %
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR