4′-Hydroxy-2-phenylacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

105980050 105980250
TS105980050EA 104 USD
TS105980050 TS105980250
4′-Hydroxy-2-phenylacetophenone 97%
4′-Hydroxy-2-phenylacetophenone
Formula: C₁₄H₁₂O₂
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00002360
CAS Number: 2491-32-9

Order Now

Specification Test Results

Appearance Cream Crystalline powder
Infrared spectrum Conforms
Melting point 147 to 151 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR