2-Amino-4-hydroxy-6-methylpyrimidine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Amino-6-methylpyrimidin-4(1H)-one, 2-Amino-6-methyl-4-pyrimidinol

103830250 103831000
TS103830250EA 33 USD
TS103830250 TS103831000
2-Amino-4-hydroxy-6-methylpyrimidine 99%
2-Amino-4-hydroxy-6-methylpyrimidine
Formula: C₅H₇N₃O
MW: 125.13 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006095
CAS Number: 3977-29-5
EINECS: 223-612-1

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
Clarity of solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR