2-Acetylfuran 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Furyl methyl ketone, 1-(Furan-2-yl)ethanone

102551000 102552500
TS102551000EA 69 USD
TS102551000 TS102552500
2-Acetylfuran 99%
2-Acetylfuran
Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 67 °C (10 mmHg)
Melting Pt: 29…30 °C
Density: 1.098 g/cm³
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003242
CAS Number: 1192-62-7
EINECS: 214-757-1
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.5060 to 1.5080 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR