Acetohydrazide 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetic hydrazide, Acethydrazide

102350250 102351000 102355000
TS102350250EA 69 USD
TS102350250 TS102351000 TS102355000
Acetohydrazide 95%
Acetohydrazide
Formula: C₂H₆N₂O
MW: 74.08 g/mol
Boiling Pt: 129 °C (18 mmHg)
Melting Pt: 57…60 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007610
CAS Number: 1068-57-1
EINECS: 213-948-7
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Appearance White Moist crystals and/or chunks
Infrared spectrum Conforms
Assay ≥94 % (on dry substance)
Loss on drying (50°C, 3 h) Balance
Gel strength ≥750 g/cm² (1 % gel)
Gel point 34.5 to 37.5°C (1.5 % gel)
Electroendosmosis 0.16 to 0.19 (-Mr)
Ash ≤0.5 %
Sulfate (SO4) ≤0.25 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR