Ceresin wax, white

Supplier: Thermo Scientific Chemicals

473452500 473450010
TS473452500EA 51 USD
TS473452500 TS473450010
Ceresin wax, white
Ceresin wax
MDL Number: MFCD00146361
CAS Number: 8001-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR