Potassium disulphate ≥95%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium disulphate (Potassium pyrosulphate), Dipotassium disulphate, Potassium pyrosulphate

472460025 472465000
TS472460025EA 260 USD
TS472460025 TS472465000
Potassium disulphate ≥95%, pure
Potassium disulphate
Formula: K₂O₇S₂
MW: 254.32 g/mol
Melting Pt: 325
MDL Number: MFCD00011385
CAS Number: 7790-62-7
UN: 2923
Merck Index: 13,07749

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR