Indole-3-butyric acid potassium salt ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium 4-(1H-indol-3-yl)butanoate

471050010 471050050 471050250
TS471050010EA 37 USD
TS471050010 TS471050050 TS471050250
Indole-3-butyric acid potassium salt ≥98%
Indole-3-butyric acid potassium salt
Formula: C₁₂H₁₂KNO₂
MW: 241.331 g/mol
MDL Number: MFCD00058442
CAS Number: 60096-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR