2-Propanol 70% in water, Technical Grade

Supplier: Thermo Scientific Chemicals

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

470610050 470610250 470610010
TS470610050EA 72 USD
TS470610050 TS470610250 TS470610010
2-Propanol 70% in water, Technical Grade
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Flash Pt: 12
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
Merck Index: 13,05228

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR