Potassium indole-3-acetate ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: K-IAA, 3-Indoleacetic acid potassium salt, Heteroauxine, Indol-3-ylacetic acid potassium salt, Indole-3-acetic acid potassium salt, Potassium 3-Indoleacetate

470280010 470280050 470280250
TS470280010EA 30 USD
TS470280010 TS470280050 TS470280250
Potassium indole-3-acetate ≥98%
Potassium indole-3-acetate
Formula: C₁₀H₈KNO₂
MW: 213.28 g/mol
MDL Number: MFCD00066213
CAS Number: 2338-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR