Nizatidine

Supplier: Thermo Scientific Chemicals

Synonyms: N-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N'-methyl-2-nitro-1,1-ethenediamine, N-(2-(((2-((Dimethylamino)methyl)thiazol-4-yl)methyl)thio)ethyl)-N-methyl-2-nitroethene-1,1-diamine

468470050 468470250
TS468470050EA 102 USD
TS468470050 TS468470250
Nizatidine
Nizatidine
Formula: C₁₂H₂₁N₅O₂S₂
MW: 331.45 g/mol
MDL Number: MFCD00865660
CAS Number: 76963-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR