Levocetirizine dihydrochloride

Supplier: Thermo Scientific Chemicals

Synonyms: 2-[2-[4-[(R)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride, (R)-2-(2-(4-((4-Chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)acetic acid dihydrochloride, Levocetirizine 2HCl

467470010 467470050
TS467470010EA 56 USD
TS467470010 TS467470050
Levocetirizine dihydrochloride
Levocetirizine dihydrochloride
Formula: C₂₁H₂₅ClN₂O₃
MW: 461.81 g/mol
MDL Number: MFCD07366507
CAS Number: 130018-87-0
UN: 3077

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR