2-Fluoro-4-methoxyphenylboronic acid ≥97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Fluoro-4-methoxybenzeneboronic acid

449840010 449840050
TS449840010EA 61 USD
TS449840010 TS449840050
2-Fluoro-4-methoxyphenylboronic acid ≥97%
2-Fluoro-4-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD04039323
CAS Number: 162101-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR