Silver nitrate 99+%, meets analytical specifications of Reagent Ph. Eur.

Supplier: Thermo Scientific Chemicals

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

194050500 194052500
TS194050500EA 269 USD
TS194050500 TS194052500
Silver nitrate 99+%, meets analytical specifications of Reagent Ph. Eur.
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
Merck Index: 12,08661

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR