Petroleum spirit, 40…60 °C, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

124860025 124860250
TS124860025EA 128 USD
TS124860025 TS124860250
Petroleum spirit, 40…60 °C, extra pure
Petroleum spirit
Boiling Pt: 40…60 °C (760 mmHg)
Density: 0.65 g/cm³
Flash Pt: –55 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.03 % (K.F.)
Boiling point 40 to 60 °C
Density (20 °C) 0.64 to 0.66 g/ml
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR