n-Heptane 99+%, pure

Supplier: Thermo Scientific Chemicals
Danger

120340010 120340025 120340050 120340251
TS120340010EA 110 USD
TS120340010 TS120340025 TS120340050 TS120340251
n-Heptane 99+%, pure
n-Heptane
Formula: H₃C(CH₂)₅CH₃
Boiling Pt: 98 °C (760 mmHg)
Melting Pt: –91 °C
Density: 0.683 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3850 to 1.3890 (20°C, 589 nm)
GC ≥99.0 %
Specific optical rotation -12° to -16° (20°C, 589 nm) (c=1% ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR