9-BBN monomer 0.5 M in THF, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

165811000 165818000
TS165811000EA 67 USD
TS165811000 TS165818000
9-BBN monomer 0.5 M in THF, AcroSeal®
9-BBN monomer
Formula: C₈H₁₅B
Density: 0.884 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00074742
CAS Number: 280-64-8
UN: 3399
ADR: 4.3,II

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid (solution under an inert atmosphere)
Strength 0.45 to 0.55 M (calc. molarity of 9-BBN based on active hydrogen)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR