Pyridine hydrofluoride 65 - 70% approx. 30% Pyridine

Supplier: Thermo Scientific Chemicals
Danger

180070100 180070500 180071000
TS180070100EA 42 USD
TS180070100 TS180070500 TS180071000
Pyridine hydrofluoride 65 - 70% approx. 30% Pyridine
Pyridine hydrofluoride

Appearance: Clear light yellow to light brown Liquid

Formula: C₅H₆FN
Density: 1.1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00012436
CAS Number: 32001-55-1
UN: 1790
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear light yellow to light brown Liquid
Titration with NaOH 65 to 70 % (HF)
Melting point 150 to 154 °C
Loss on drying ≤0.5 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR