Potassium-tert-pentyloxide 14% (w/w) 0.9 M in cyclohexane, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium tert-amylate, Potassium 2-methyl-2-butoxide, Potassium tert-pentoxide

164721000 164728000
TS164721000EA 67 USD
TS164721000 TS164728000
Potassium-tert-pentyloxide 14% (w/w) 0.9 M in cyclohexane, AcroSeal®
Potassium-tert-pentyloxide
Formula: C₅H₁₁KO
MW: 126.24 g/mol
Density: 0.82 g/cm³
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00064808
CAS Number: 41233-93-6
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to yellow Solution
Titration with HClO4 0.85 to 1.10 M
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR