Nerolidol (cis and trans mixture) 97% (total alcohols)

Supplier: Thermo Scientific Chemicals
Warning

121970250 121971000
TS121970250EA 36 USD
TS121970250 TS121971000
Nerolidol (cis and trans mixture) 97% (total alcohols)
Nerolidol (cis and trans mixture)
Formula: C₁₅H₂₆O
Boiling Pt: 114 °C (1 mmHg)
Melting Pt: –75 °C
Density: 0.87 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00008911
CAS Number: 7212-44-4
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4830 (20 °C, 589 nm)
GC ≥97.0 % (total alcohols)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR