Sodium deuteroxide 40% (w/w) in D₂O for NMR spectroscopy (D₂O 99+ atom%D)

Supplier: Thermo Scientific Chemicals
Danger

175470100 175470500
TS175470100EA 42 USD
TS175470100 TS175470500
Sodium deuteroxide 40% (w/w) in D₂O for NMR spectroscopy (D₂O 99+ atom%D)
Sodium deuteroxide
Formula: NaOD
Density: 1.46 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00037669
CAS Number: 14014-06-3
EINECS: 237-825-2
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
D-Enrichment ≥99.0 %
Concentration 40 to 42 % w/w
HPLC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR