Sulfuric acid-[D₂] 98% (w/w) in D₂O for NMR spectroscopy (D₂O 99.5+ atom%D)
Supplier: Thermo Scientific Chemicals
|
Danger
|
175480050
175480500
175481000
TS175480050EA
28
USD
TS175480050
TS175480500
TS175481000
Sulfuric acid-[D₂] 98% (w/w) in D₂O for NMR spectroscopy (D₂O 99.5+ atom%D)
Sulfuric acid-[D₂]
Formula:
D₂SO₄ Density: 1.87 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003542 CAS Number: 13813-19-9 UN: 1830 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless Solution |
D-Enrichment | ≥99.50 % |
Titration with NaOH | 97.5 to 102.5 % |
GC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...