Triethyloxonium tetrafluoroborate 1 M in methylene chloride

Supplier: Thermo Scientific Chemicals
Danger

175110250 175111000
TS175110250EA 67 USD
TS175110250 TS175111000
Triethyloxonium tetrafluoroborate 1 M in methylene chloride
Triethyloxonium tetrafluoroborate
Formula: C₆H₁₅BF₄O
Density: 1.328 g/cm³
Storage Temperature: Freezer
MDL Number: MFCD00044423
CAS Number: 368-39-8
EINECS: 206-705-1
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Solution
Strength 0.9 to 1.1 M
Melting point 148 to 153 °C
Loss on drying ≤0.5 % (1 g, 105 °C, 1 h)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR