Propagyl bromide 80% (w/w) in toluene stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Bromopropyne

131480010 131480500 131482500
TS131480010EA 716 USD
TS131480010 TS131480500 TS131482500
Propagyl bromide 80% (w/w) in toluene stabilised
Propagyl bromide
Formula: C₃H₃Br
MW: 118.96 g/mol
Boiling Pt: 90
Melting Pt: -43
Density: 1.38 g/cm³
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000241
CAS Number: 106-96-7
EINECS: 203-447-1
UN: 3286
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to yellow-orange Solution
Refractive index 1.4910 to 1.4970 (20°C, 589 nm)
Titration Argentometric 75 to 85 % (Product in toluene)
Density (20°C) 1.33 to 1.38 g/ml
GC ≥96.0 % (Excluding toluene)
Stabilizer 0.3 % MgO


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR