Styrene-[D₈] 98+ atom% D stabilized for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
175390010
175390050
TS175390010EA
107
USD
TS175390010
TS175390050
Styrene-[D₈] 98+ atom% D stabilized for NMR spectroscopy
Styrene-[D₈]
Formula:
C₆D₅CDCD₂ Boiling Pt: 50…51 °C (25 mmHg) Melting Pt: –31 °C Density: 0.97 g/cm³ Flash Pt: 32 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00044231 CAS Number: 19361-62-7 EINECS: 242-995-6 UN: 2055 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.05 % (Coulometric) |
Stabilizer | 0.5 % 4-tert-Butylcatechol |
D-Enrichment | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...